Simpson 260-8 Analog Series Multimeter
Accuracy for the lab, rugged construction for the field
  • Excellent for adjustments such as nulling and peaking
  • Large color code scale
  • AC frequency response to 100 KHz
  • Zero RFI
  • Standard and high energy overload fusing
  • Varistor protected meter movement
  • Mirrored scale
  • Broad range of accessories and probes
  • Wide range capabilities
  • Overload protection relay circuits
The Simpson 260 analog meter is ideal when monitoring fluctuating trends and rates. It features more attributes than its predecessors. Standard and high energy fusing in conjunction with diode network meter movement protection makes
the meter the best combination of quality and affordability. An additional relay overload protection circuit is resettable and provides audible continuity checking.
117,000.00 - ₹191,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Simpson 260-8  12388 Black Analog Multimeter WW-20006-06 Mfr # 12388
COMPARE
Black Analog Multimeter
₹117,000.00INR / Each
Simpson 260-8P  12391 Relay Protected Black Analog Multimeter WW-20006-07 Mfr # 12391
COMPARE
Relay Protected Black Analog Multimeter
₹141,000.00INR / Each
Simpson 260-8XI  12395 Yellow Analog Multimeter WW-20006-08 Mfr # 12395
COMPARE
Yellow Analog Multimeter
₹124,000.00INR / Each
Simpson 260-8XPI 12396 Relay Protected Yellow Analog Multimeter WW-20006-11 Mfr # 12396
COMPARE
Relay Protected Yellow Analog Multimeter
₹164,000.00INR / Each
Simpson VOMLOW POWER PROTECT WW-20023-66 Mfr # 12211
COMPARE
₹191,000.00INR / Each