Silicone Self-fusing Tape
Self-bonding, self-fusing silicone tape resists moisture, oxygen, and ozone. Highly resistant to voltages-provides a long service life. Use this cohesive insulating barrier from -150 to 500°F (-101 to 260°C). Fuses to itself in about 24 hours, forming an inseparable bond. This flat tape has no adhesives and leaves no residue. Protects from effects of vibration and shock.
5,600.00 - ₹5,700.00INR / Roll of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (ft) Thickness (mil)
Availability Pricing
COMPARE
Orange 108 0.5
₹5,600.00INR / Roll of 1
COMPARE
Black 108 0.5
₹5,700.00INR / Roll of 1
COMPARE
White 108 0.5
₹5,700.00INR / Roll of 1
COMPARE
Blue 108 0.5
₹5,700.00INR / Roll of 1