BriskHeat® Silicone Rubber Heating Tapes with Adjustable Time Percentage Control
Regulates the length of time the heater is on and off
  • Heat to 500°F (260°C)
  • Flexible silicone rubber is moisture resistant
Solve a wide range of heating applications without risk of damage due to chemical and
moisture spillover. Tapes are available with either built-in time percentage control. Tapes are made of fine-strand resistance wires that are double insulated with braided fiberglass and knitted into flat tapes; then completely enclosed in highstrength, high-temperature-resistant silicone rubber. These tapes are ideal for tight radius heating applications and are chemical, abrasion and moisture resistant.
Warning: Heating tapes should not be overlapped.
28,200.00 - ₹47,500.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
BriskHeat 51002 Silicone Heating Tape with Adjustable Control, 0.5" W x 2'L, 72W; 120V with Plug 03106-32 WW-03106-32 Mfr # TBSAT051-020
COMPARE
450 120 Silicone Heating Tape with Adjustable Control, 0.5\" W x 2'L, 72W; 120V with Plug 03106-32
₹28,800.00INR / Each
BriskHeat 51004 Silicone Heating Tape with Adjustable Control, 0.5" W x 4'L, 144W; 120V with Plug 03106-33 WW-03106-33 Mfr # TBSAT051-040
COMPARE
450 120 Silicone Heating Tape with Adjustable Control, 0.5\" W x 4'L, 144W; 120V with Plug 03106-33
₹28,200.00INR / Each
BriskHeat 51006 Silicone Heating Tape with Adjustable Control, 0.5" W x 6'L, 216W; 120V with Plug WW-03106-34 Mfr # TBSAT051-060
COMPARE
450 120 Silicone Heating Tape with Adjustable Control, 0.5\" W x 6'L, 216W; 120V with Plug
₹29,600.00INR / Each
BriskHeat 51008 Silicone Heating Tape with Adjustable Control, 0.5" W x 8'L, 288W; 120V with Plug WW-03106-35 Mfr # TBSAT051-080
COMPARE
450 120 Silicone Heating Tape with Adjustable Control, 0.5\" W x 8'L, 288W; 120V with Plug
₹32,900.00INR / Each
BriskHeat 51010 Silicone Heating Tape with Adjustable Control, 0.5" W x 10'L, 360W; 120V with Plug WW-03106-36 Mfr # TBSAT051-100
COMPARE
450 120 Silicone Heating Tape with Adjustable Control, 0.5\" W x 10'L, 360W; 120V with Plug
₹34,300.00INR / Each
BriskHeat TBSAT101-020 Silicone Heating Tape with Controller (2' L x 1" W) WW-36001-52 Mfr # TBSAT101-020
COMPARE
500 120 Silicone Heating Tape with Controller (2' L x 1\" W)
₹31,000.00INR / Each
BriskHeat TBSAT101-040 Silicone Heating Tape with Controller (4' L x 1" W) WW-36001-54 Mfr # TBSAT101-040
COMPARE
500 120 Silicone Heating Tape with Controller (4' L x 1\" W)
₹33,500.00INR / Each
BriskHeat TBSAT101-060 Silicone Heating Tape with Controller (6' L x 1" W) WW-36001-56 Mfr # TBSAT101-060
COMPARE
500 120 Silicone Heating Tape with Controller (6' L x 1\" W)
₹41,700.00INR / Each
BriskHeat TBSAT101-080 Silicone Heating Tape with Controller (8' L x 1" W) WW-36001-58 Mfr # TBSAT101-080
COMPARE
500 120 Silicone Heating Tape with Controller (8' L x 1\" W)
₹42,700.00INR / Each
BriskHeat TBSAT101-100 Silicone Heating Tape with Controller (10' L x 1" W) WW-36001-60 Mfr # TBSAT101-100
COMPARE
450 120 Silicone Heating Tape with Controller (10' L x 1\" W)
₹47,500.00INR / Each
Showing 1- 10 of 10