BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces with Control and Adhesive-120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 160°F (71°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 160°F . No additional control is necessary. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface.
37,900.00 - ₹144,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06121PADJB Silicone Heating Blanket with Controller, 50-160F, 6x12 Size, 120 Volt, 90 Watt WW-36002-52 Mfr # SRP06121PADJB
COMPARE
12 160 120
₹37,900.00INR / Each
BriskHeat SRP06241PADJB Silicone Heating Blanket with Controller, 50-160F, 6x24 Size, 120 Volt, 180 Watt WW-36002-54 Mfr # SRP06241PADJB
COMPARE
24 160 120
₹42,800.00INR / Each
BriskHeat SRP06361PADJB Silicone Heating Blanket with Controller, 50-160F, 6x36 Size, 120 Volt, 270 Watt WW-36002-56 Mfr # SRP06361PADJB
COMPARE
36 160 120
₹59,200.00INR / Each
BriskHeat SRP12121PADJB Silicone Heating Blanket with Controller, 50-160F, 12x12 Size, 120 Volt, 180 Watt WW-36002-58 Mfr # SRP12121PADJB
COMPARE
12 160 120
₹43,300.00INR / Each
BriskHeat SRP12241PADJB Silicone Heating Blanket with Controller, 50-160F, 12x24 Size, 120 Volt, 360 Watt WW-36002-60 Mfr # SRP12241PADJB
COMPARE
24 160 120
₹64,500.00INR / Each
BriskHeat SRP12361PADJB Silicone Heating Blanket with Controller, 50-160F, 12x36 Size, 120 Volt, 540 Watt WW-36002-62 Mfr # SRP12361PADJB
COMPARE
36 160 120
₹98,300.00INR / Each
BriskHeat SRP18361PADJB Silicone Heating Blanket with Controller, 50-160F, 18x36 Size, 120 Volt, 810 Watt WW-36002-66 Mfr # SRP18361PADJB
COMPARE
36 160 120
₹136,000.00INR / Each
BriskHeat SRP24241PADJB Silicone Heating Blanket with Controller, 50-160F, 24x24 Size, 120 Volt, 720 Watt WW-36002-68 Mfr # SRP24241PADJB
COMPARE
24 160 120
₹112,000.00INR / Each
BriskHeat SRP24361PADJB Silicone Heating Blanket with Controller, 50-160F, 24x36 Size, 120 Volt, 1080 Watt WW-36002-70 Mfr # SRP24361PADJB
COMPARE
36 160 120
₹144,000.00INR / Each
Showing 1- 9 of 9