BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces with Control-120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 160°F (71°C
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 160°F . No additional control is necessary. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface.
33,200.00 - ₹125,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06121ADJB Silicone Heating Blanket with Controller, 90 W, for Plastics, 6" x 12" , 120 V WW-36002-72 Mfr # SRP06121ADJB
COMPARE
12 160 120
₹33,200.00INR / Each
BriskHeat SRP06241ADJB Silicone Heating Blanket with Controller, 180 W, for Plastics, 6" x 24" , 120 V WW-36002-74 Mfr # SRP06241ADJB
COMPARE
24 160 120
₹40,800.00INR / Each
BriskHeat SRP06361ADJB Silicone Heating Blanket with Controller, 270 W, for Plastics, 6" x 36" , 120 V WW-36002-76 Mfr # SRP06361ADJB
COMPARE
36 160 120
₹53,900.00INR / Each
BriskHeat SRP12121ADJB Silicone Heating Blanket with Controller, 180 W, for Plastics, 12" x 12" , 120 V WW-36002-78 Mfr # SRP12121ADJB
COMPARE
12 160 120
₹40,400.00INR / Each
BriskHeat SRP12241ADJB Silicone Heating Blanket with Controller, 360 W, for Plastics, 12" x 24" , 120 V WW-36002-80 Mfr # SRP12241ADJB
COMPARE
24 160 120
₹58,400.00INR / Each
BriskHeat SRP12361ADJB Silicone Heating Blanket with Controller, 540 W, for Plastics, 12" x 36" , 120 V WW-36002-82 Mfr # SRP12361ADJB
COMPARE
36 160 120
₹87,400.00INR / Each
BriskHeat SRP18361ADJB Silicone Heating Blanket with Controller, 5810 W, for Plastics, 18" x 36" , 120 V WW-36002-86 Mfr # SRP18361ADJB
COMPARE
36 160 120
₹122,000.00INR / Each
BriskHeat SRP24361ADJB Silicone Heating Blanket with Controller, 1080 W, for Plastics, 24" x 36" , 120 V WW-36002-90 Mfr # SRP24361ADJB
COMPARE
36 160 120
₹125,000.00INR / Each
Showing 1- 8 of 8