BriskHeat® Silicone-Rubber Heater Blankets Plastic Surfaces with Adhesive-120 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Models for plastic surfaces have a lower 1.25 watts/in2 power density to prevent scorching of the surface.
21,800.00 - ₹83,200.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRP06121P Silicone Heating Blanket, 6x12 Size, 120 Volt, 90 Watt WW-36001-53 Mfr # SRP06121P
COMPARE
12 450 120
₹21,800.00INR / Each
BriskHeat SRP06241P Silicone Heating Blanket, 6x24 Size, 120 Volt, 180 Watt WW-36001-57 Mfr # SRP06241P
COMPARE
24 450 120
₹30,600.00INR / Each
BriskHeat SRP06361P Silicone Heating Blanket, 6x36 Size, 120 Volt, 270 Watt WW-36001-62 Mfr # SRP06361P
COMPARE
36 450 120
₹44,000.00INR / Each
BriskHeat SRP12121P Silicone Heating Blanket, 12x12 Size, 120 Volt, 180 Watt WW-36001-64 Mfr # SRP12121P
COMPARE
12 450 120
₹27,700.00INR / Each
BriskHeat SRP12241P Silicone Heating Blanket, 12x24 Size, 120 Volt, 360 Watt WW-36001-66 Mfr # SRP12241P
COMPARE
24 450 120
₹49,400.00INR / Each
BriskHeat SRP12361P Silicone Heating Blanket, 12x36 Size, 120 Volt, 540 Watt WW-36001-68 Mfr # SRP12361P
COMPARE
36 450 120
₹83,200.00INR / Each