BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces with Control and Adhesive-240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 425°F (218°C)
  • Easy-to-apply industrial strength pressure sensitive adhesive backing
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications.
Blanket with Control offers extra convenience and functionality with its included adjustable thermostat control. Models for metal surfaces can be adjusted between 50 to 425°F. No additional control is necessary
The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
37,900.00 - ₹144,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06122PADJB Silicone Heating Blanket with Controller, 50-425F, 6x12 Size, 240 Volt, 180 Watt WW-36001-99 Mfr # SRL06122PADJB
COMPARE
12 425 240
₹37,900.00INR / Each
BriskHeat SRL06242PADJB Silicone Heating Blanket with Controller, 50-425F, 6x24 Size, 240 Volt, 360 Watt WW-36002-03 Mfr # SRL06242PADJB
COMPARE
24 425 240
₹42,800.00INR / Each
BriskHeat SRL06362PADJB Silicone Heating Blanket with Controller, 50-425F, 6x36 Size, 240 Volt, 540 Watt WW-36002-07 Mfr # SRL06362PADJB
COMPARE
36 425 240
₹59,200.00INR / Each
BriskHeat SRL12122PADJB Silicone Heating Blanket with Controller, 50-425F, 12x12 Size, 240 Volt, 360 Watt WW-36002-11 Mfr # SRL12122PADJB
COMPARE
12 425 240
₹43,300.00INR / Each
BriskHeat SRL12242PADJB Silicone Heating Blanket with Controller, 50-425F, 12x24 Size, 240 Volt, 720 Watt WW-36002-15 Mfr # SRL12242PADJB
COMPARE
24 425 240
₹64,500.00INR / Each
BriskHeat SRL18182PADJB Silicone Heating Blanket with Controller, 50-425F, 18x18 Size, 240 Volt, 810 Watt WW-36002-23 Mfr # SRL18182PADJB
COMPARE
18 425 240
₹78,000.00INR / Each
BriskHeat SRL18362PADJB Silicone Heating Blanket with Controller, 50-425F, 18x36 Size, 240 Volt, 1472 Watt WW-36002-27 Mfr # SRL18362PADJB
COMPARE
36 425 240
₹136,000.00INR / Each
BriskHeat SRL24242PADJB Silicone Heating Blanket with Controller, 50-425F, 24x24 Size, 240 Volt, 1440 Watt WW-36002-31 Mfr # SRL24242PADJB
COMPARE
24 425 240
₹112,000.00INR / Each
BriskHeat SRL24362PADJB Silicone Heating Blanket with Controller, 50-425F, 12x36 Size, 240 Volt, 2160 Watt WW-36002-32 Mfr # SRL24362PADJB
COMPARE
36 425 240
₹144,000.00INR / Each
Showing 1- 9 of 9