BriskHeat® Silicone-Rubber Heater Blankets Metal Surfaces - 240 VAC
Rugged, moisture / chemical resistant flexible heaters provide uniform heat distribution across large surface areas
  • Maximum exposure temperature of 450°F (232°C)
These versatile and industrial-strength heaters can be used on a wide range of surfaces and environments due to their flexibility, moisture / chemical resistance, and ruggedness. They easily install on everything from laboratory vessels to large vats and tanks - ideal for solving viscosity, freeze protection, and process control heating applications. The unique grounded heating element is laminated between two layers of extra strong 20mil fiberglass reinforced silicone rubber. Heaters for metal surfaces have faster heat-up rates due to the 2.5 watts /in2 power density.
25,400.00 - ₹110,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat SRL06242 Silicone Heating Blanket, 6x24 Size, 240 Volt, 360 Watt, for metal surfaces WW-36001-29 Mfr # SRL06242
COMPARE
24 450 240
₹27,800.00INR / Each
BriskHeat SRL12122 Silicone Heating Blanket, 12x12 Size, 240 Volt, 360 Watt, for metal surfaces WW-36001-34 Mfr # SRL12122
COMPARE
12 450 240
₹25,400.00INR / Each
BriskHeat SRL12242 Silicone Heating Blanket, 12x24 Size, 240 Volt, 720 Watt, for metal surfaces WW-36001-37 Mfr # SRL12242
COMPARE
24 450 240
₹43,600.00INR / Each
BriskHeat SRL24362 Silicone Heating Blanket, 24x36 Size, 240 Volt, 2160 Watt, for metal surfaces WW-36001-51 Mfr # SRL24362
COMPARE
36 450 240
₹110,000.00INR / Each