This product is now discontinued, View Related Products

Signs; Flammable - No Smoking

These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. They are intended to define specific hazards by conveying a clear, easily understood message. The sign headers, and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.

Signs are available in Adhesive Vinyl (VS), Plastic (VP), and Aluminum (VA) to best suit your application. VS is a flexible 4-mil pressure sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low cost, light duty applications. VP is .055” polyethylene plastic and is a good medium duty indoor and sheltered outdoor environments. VA is .040” aluminum with over laminate
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MSMK968VS Safety Sign, Flammable - No Smoking, 7" X 10", Adhesive Vinyl WW-60710-28 Mfr # MSMK968VS
COMPARE
7 Vinyl 10
Call For Price
Accuform MSMK968VP Safety Sign, Flammable - No Smoking, 7" X 10", Plastic WW-60710-29 Mfr # MSMK968VP
COMPARE
7 Plastic 10
Call For Price
Accuform MSMK968VA Safety Sign, Flammable - No Smoking, 7" X 10", Aluminum WW-60710-30 Mfr # MSMK968VA
COMPARE
7 Aluminum 10
Call For Price
Accuform MSMKG49VS Safety Sign, Flammable - No Smoking, 10" X 14", Adhesive Vinyl WW-61014-28 Mfr # MSMKG49VS
COMPARE
10 Vinyl 14
Call For Price
Accuform MSMKG49VP Safety Sign, Flammable - No Smoking, 10" X 14", Plastic WW-61014-29 Mfr # MSMKG49VP
COMPARE
10 Plastic 14
Call For Price
Accuform MSMKG49VA Safety Sign, Flammable - No Smoking, 10" X 14", Aluminum WW-61014-30 Mfr # MSMKG49VA
COMPARE
10 Aluminum 14
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat