Signs; Emergency Exit Only (red/white)
These signs are intended to indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. They are intended to define specific hazards by conveying a clear, easily understood message. The sign headers, and colors used in accident prevention safety signs are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
Signs are available in Adhesive Vinyl (VS), Plastic (VP), and Aluminum (VA) to best suit your application. VS is a flexible 4-mil pressure sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low cost, light duty applications. VP is .055” polyethylene plastic and is a good medium duty indoor and sheltered outdoor environments. VA is .040” aluminum with over laminate
2,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MEXT584VP Safety Sign, Emergency Exit Only (red/white), 7 X 10, Plastic WW-60710-44 Mfr # MEXT584VP
COMPARE
7 Plastic 10
₹2,100.00INR / Each