WS Tyler Sieve Covers and Receiving Pans for 8 inch Diameter Sieves
Protect and collect samples easily
  • Suitable for use with 8” diameter sieves
  • Constructed of durable stainless steel or brass
Use covers and receiving pans with 8” diameter sieves. Order sieves separately. Order a receiving pan for the bottom sieve to collect the finest particles. Receiving pans with a fitted rim for nesting allow you to collect from between sieves, not just the bottom sieve.
6,000.00 - ₹15,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Outside Diameter (in) Description
Availability Pricing
W.S. Tyler 8481 Receiving Pan for Bottom 8" Diameter Brass Sieve, Full Height WW-59986-80 Mfr # 8481
COMPARE
Brass 8 Receiving Pan for Bottom 8\" Diameter Brass Sieve, Full Height
₹6,000.00INR / Each
W.S. Tyler 8482 Receiving Pan for Bottom 8" Diameter Brass Sieve, Half Height WW-59986-81 Mfr # 8482
COMPARE
Brass 8 Receiving Pan for Bottom 8\" Diameter Brass Sieve, Half Height
₹6,000.00INR / Each
W.S. Tyler 8483 Receiving Pan with Fitted Rim for Nesting 8" Full Height Brass Sieves WW-59986-82 Mfr # 8483
COMPARE
Brass 8 Receiving Pan with Fitted Rim for Nesting 8\" Full Height Brass Sieves
₹10,000.00INR / Each
W.S. Tyler 8484 Receiving Pan with Fitted Rim for Nesting 8" Half Height Brass Sieves WW-59986-83 Mfr # 8484
COMPARE
Brass 8 Receiving Pan with Fitted Rim for Nesting 8\" Half Height Brass Sieves
₹10,300.00INR / Each
W.S. Tyler 8488 Top Cover with Handle for 8" Diameter Brass Sieve WW-59986-85 Mfr # 8488
COMPARE
Brass 8 Top Cover with Handle for 8\" Diameter Brass Sieve
₹6,900.00INR / Each
W.S. Tyler 8491 Receiving Pan for 8" Diameter Stainless Steel Full Height Sieves WW-59986-90 Mfr # 8491
COMPARE
Stainless Steel 8 Receiving Pan for 8\" Diameter Stainless Steel Full Height Sieves
₹10,900.00INR / Each
W.S. Tyler 8492 Receiving Pan for 8" Diameter Stainless Steel Half Height Sieves WW-59986-91 Mfr # 8492
COMPARE
Stainless Steel 8 Receiving Pan for 8\" Diameter Stainless Steel Half Height Sieves
₹10,900.00INR / Each
W.S. Tyler 8493 Receiving Pan with Fitted Rim for Nesting 8" Stainless Steel Sieves, Full Height WW-59986-92 Mfr # 8493
COMPARE
Stainless Steel 8 Receiving Pan with Fitted Rim for Nesting 8\" Stainless Steel Sieves, Full Height
₹13,200.00INR / Each
W.S. Tyler 8494 Receiving Pan with Fitted Rim for Nesting 8" Stainless Steel Sieves, Half Height WW-59986-93 Mfr # 8494
COMPARE
Stainless Steel 8 Receiving Pan with Fitted Rim for Nesting 8\" Stainless Steel Sieves, Half Height
₹13,200.00INR / Each
W.S. Tyler 8497 Top Cover with Handle for 8" Stainless Steel Sieve WW-59986-95 Mfr # 8497
COMPARE
Stainless Steel 8 Top Cover with Handle for 8\" Stainless Steel Sieve
₹15,000.00INR / Each
Showing 1- 10 of 10