Cole-Parmer® Essentials Sieve Covers and Receiving Pans for 3" Diameter Sieves
Protect and collect samples easily
  • Suitable for use with 3” diameter sieves
  • Constructed of durable stainless steel
Use covers and receiving pans with 3” diameter sieves. Order sieves separately. Order a receiving pan for the bottom sieve to collect the finest particles.
5,800.00 - ₹20,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
3\" Diameter Stainless Steel Sieve Cover
₹5,800.00INR / Each
COMPARE
3\" Diameter Stainless Steel Full Height Sieve Pan
₹20,000.00INR / Each