Shortwave Ultraviolet Lamp

Enhance the performance of your imagine system
  • Compatible with MultiDoc-It™ Multi-User Imaging System
  • Two lamps are recommended for uniform illumination
When overhead UV illumination is required in your MultiDoc-It™ Multi-Use Imaging System, UV lamps are required. Two lamps are recommended for uniform illumination. Choose the lamp based off of the wavelength your application requires
44,200.00 - ₹66,200.00INR / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Description
Availability Pricing
Analytik Jena 95-0016-15 Shortwave UV lamp; 4 watts, 230 VAC/50 Hz WW-09815-02 Mfr # 95-0016-15
COMPARE
50 230 Shortwave UV lamp; 4 watts, 230 VAC/50 Hz
₹66,200.00INR / Each
Analytik Jena UVL-21 UV Lamp, 4-Watt, 365 nm; 230 VAC WW-09817-02 Mfr # 95-0018-03
COMPARE
50 230 UV Lamp, 4-Watt, 365 nm; 230 VAC
₹44,200.00INR / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat