This product is now discontinued, View Related Products
Shimpo TRC Series Digital Torque Checkers
Fast, accurate, and convenient checking of torque wrenches, torque drivers, and torque watches.
  • User selectable units of measurement
  • LED’s with peak display and memory recalls, clockwise, counterclockwise
  • Includes Digi-Torq software for calibration and data analysis capabilities
  • USB output capabilities
  • Units store up to 10 test set-ups
  • 3 units of measurement - 2Nm, 5Nm, 10Nm
  • Battery or AC operation for additional flexibility
The Shimpo TRC-2, TRC-5, and TRC-10 digital torque meters are easy to set up and use with the push a single button. Simply zero, test, then store. Audible alarms and visual (LED’s) indicators alert the operator of high/no-go, go, and low/no-go conditions. Meters store up to 3000 test data points and included DigiTorq software to help track all your torque driver calibrations and provide powerful data analysis capabilities. The heavy duty cast aluminum meters are designed with a mounting bracket and removable rubber feet for permanent mounting on a cart or bench. The compact design is extremely durable and ideal for applications on the plant floor or out in the field.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m)
Availability Pricing
Shimpo TRC-2 Model Digital Torque Tester WW-59891-21 Mfr # TRC-2
COMPARE
17.7 2
Call For Price
Shimpo TRC-5 Model Digital Torque Tester WW-59891-23 Mfr # TRC-5
COMPARE
44.25 5
Call For Price
Shimpo TRC-10 Model Digital Torque Tester WW-59891-25 Mfr # TRC-10
COMPARE
88.5 10