This product is now discontinued, View Related Products
Servus® by Honeywell™ 12” H Waterproof Neoprene Steel-Toed Work Boots
Keep workers’ feet dry and comfortable
  • Triple dipped in neoprene for 100% waterproof boots
  • Chemical-resistant neoprene construction
  • Electrical hazard rated outsole and steel toe impact protection
  • Meet ASTM F2413-18 M/I/C EH requirements
  • Available in mens' sizes 4 to 13
Servus® by Honeywell™ work boots feature hand-laid natural rubber to offer flexibility and strength at critical stress points. The boots are then triple dipped in neoprene and vulcanized to create a 100% seamless, liquid-tight boot. Self-cleaning chevron outer sole has a flat, surface-to-surface tread pattern for increased slip resistance on slick surfaces and the open channels keep out liquids and debris. The work boots meet ASTM F2413-18 hazard and steel toe (M 1/75 & C75) requirements. The removable and washable Breath-O-Prene® liner absorbs shocks and keeps away moisture to keep your feet dry and comfortable.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Men's Size
Availability Pricing
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 4; 1 Pair WW-86326-78 Mfr # 22114-CTM-040
COMPARE
12 Neoprene 4
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 5; 1 Pair WW-86326-79 Mfr # 22114-CTM-050
COMPARE
12 Neoprene 5
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 6; 1 Pair WW-86326-80 Mfr # 22114-CTM-060
COMPARE
12 Neoprene 6
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 7; 1 Pair WW-86326-81 Mfr # 22114-CTM-070
COMPARE
12 Neoprene 7
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 8; 1 Pair WW-86326-82 Mfr # 22114-CTM-080
COMPARE
12 Neoprene 8
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 9; 1 Pair WW-86326-83 Mfr # 22114-CTM-090
COMPARE
12 Neoprene 9
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 10; 1 Pair WW-86326-84 Mfr # 22114-CTM-100
COMPARE
12 Neoprene 10
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 11; 1 Pair WW-86326-85 Mfr # 22114-CTM-110
COMPARE
12 Neoprene 11
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 12; 1 Pair WW-86326-86 Mfr # 22114-CTM-120
COMPARE
12 Neoprene 12
Call For Price
Honeywell Servus Neoprene Steel Toe Men's Work Boots, Size 13; 1 Pair WW-86326-87 Mfr # 22114-CTM-130
COMPARE
12 Neoprene 13
Call For Price
Showing 1- 10 of 10