BTO® Bleed and Temperature Optimized Septa
Optimized for the most demanding GC and GC/MS applications
  • Formulated to extend low-bleed and outstanding mechanical properties
BTO® (Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.
15,100.00 - ₹16,700.00INR / Pkg of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
BTO Bleed and Temperature Optimized Septa, 3/8" (9.5 mm); 50/Pk WW-01994-30 Mfr # 298705
COMPARE
(Bleed/Temp Optimized) Septa, 3/8\" (9.5 mm); 50/Pack
₹15,100.00INR / Pkg of 50
BTO Bleed and Temperature Optimized Septa, CenterGuide, 7/16" (11 mm); 50/Pk WW-01994-32 Mfr # 298717
COMPARE
(Bleed/Temp Optimized) Septa, CenterGuide, 7/16\" (11 mm); 50/Pk
₹15,100.00INR / Pkg of 50
BTO Bleed and Temperature Optimized Septa, CenterGuide, 9 mm; 50/Pk WW-01999-10 Mfr # 298713
COMPARE
(Bleed/Temp Optimized) Septa, CenterGuide, 9 mm; 50/Pk
₹15,100.00INR / Pkg of 50
BTO Bleed and Temperature Optimized Septa, Shimadzu; 50/Pk WW-01999-14 Mfr # 298735
COMPARE
(Bleed/Temp Optimized) Septa, Shimadzu; 50/Pk
₹16,700.00INR / Pkg of 50