This product is now discontinued, View Related Products
Self-Masking Quartz Micro Cuvette for UV Spectroscopy
  • Matched pairs for less data variability
Cuvettes come in matched pairs in standard, semimicro, or micro size, and can be ordered with either a cover of PTFE resin or a stopper of PTFE resin. All cuvettes have a 10-mm pathlength. All pairs are accurately matched to give the same value for the same solution within 1% tolerance. Quartz cuvettes can be used for the entire UV spectra down to 200 nm and have a guaranteed 80% transmission at 200 nm.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
Quartz 0.7 10