Screw-Cap and Bung-Mount IBC Tote Mixers
Easily customize these units to fit your drum requirements
  • Gear drive mixers include 9-inch folding propeller to easily mix materials in a drum
  • Available in electric or air-drive models
These mixers are perfect for mixing contents of drums and semi-bulk shipping containers. Each unit includes a 316SS shaft and propeller. The propeller folds to fit through the drum opening, and opens to 9-inches in diameter. Mixers mount directly to a 2-inch coupling or flange, or order the optional cap mounting flange to use mixer on any plastic screw cap on bulk mixing containers. Simply mount the flange to your bulk cap by drilling four holes, mixer attaches to the flange and you are ready to mix.
Mixers are available in either TEFC or air-drive versions.
460,000.00 - ₹534,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Motor hp Motor Type
Availability Pricing
Neptune Bung-Mount IBC Tote Mixer, 430 rpm, 3/4 HP; 115-230V WW-50317-05 Mfr # HGL-5.0
COMPARE
430 3/4 TEFC
₹460,000.00INR / Each
Neptune Bung-Mount IBC Tote Mixer, 430 rpm, 1 HP; 115-230V WW-50317-10 Mfr # HGL-6.0
COMPARE
430 1 TEFC
₹534,000.00INR / Each