Scientific Industries Vortex-Genie® Max Shakers
Provides high speed vortexing up to 2000 rpm
  • Ratcheting clamp system securely holds a variety of containers
  • Programmable gentle ramp up to speed allows use of open containers
  • Durable, heavy zinc die casting with specially designed feet prevents ''walking''
  • Ideal for repetitive protocols
The fully programmable, large capacity multi-vessel vortex shaker offers high speed vortexing up to 2000 rpm. Digital control and display of speed and time provide accuracy, reproducibility, and repeatability with true speed display. The specially designed clamping platform can securely hold up to six 250 mL vessels, 20 test tubes (10 per rack) of any kind up to 29 mm in dia, or four microplates.
Choose from normal, reverse, and pulse modes of operation. To set a desired cycle time from one minute to 99 hours and 59 minutes, select the Normal mode. If you wish to switch directions at regular intervals, choose the Reverse mode. To toggle between operating in one direction and stopping at regular intervals, press the Pulse mode button.
590,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
Scientific Industries Vortex-Genie® Max Shaker, 120V WW-04724-06 Mfr # SI-MX200
COMPARE
2000 120 Heavy duty Vortex Shaker, 120V
₹590,000.00INR / Each
Scientific Industries Vortex-Genie® Max Shaker, 230V w/o plug WW-04724-07 Mfr # SI-MX201
COMPARE
1950 230 Heavy duty Vortex Shaker, 230V w/o plug
₹590,000.00INR / Each
Scientific Industries Vortex-Genie® Max Shaker, 230V Euro plug WW-04724-08 Mfr # SI-MX202
COMPARE
1950 230 Heavy duty Vortex Shaker, 230V Euro plug
₹590,000.00INR / Each