This product is now discontinued, View Related Products
Scientific Industries Bag Rotator Genie
Rotator for effective mixing of sealed bags
  • Maintain sample components in suspension
  • Turn up to forty 300-mL transfer packs in an end-over-end motion—at a time
  • Runs at a fixed speed of 3 rpm to provide uniform samples
  • Gentle agitation minimizes cellular trauma and prevents clumping
  • Durable construction provides years of maintenance-free performance
This bag mixer-rotator effectively mixes samples in transfer packs or sealed bags. The weighted base stays put and won't tip over, and the brushless motor makes it maintenance-free for years of dependable use.
The included wire basket holds up to 40 standard 300-mL bags at a time. Bags are gently rotated end-over-end to ensure consistent mixing of suspensions. The fixed speed operation of 3 rpm minimizes lysis and sample disruption.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Description
Availability Pricing
Scientific Industries Bag Rotator Genie Rotating Shaker, 120V with North American Plug WW-04724-11 Mfr # SI-1000
COMPARE
50/60 120 Rotating Shaker, 120V with North American Plug
Call For Price
Scientific Industries Bag Rotator Genie Rotating Shaker, 230V without Plug WW-04724-12 Mfr # SI-1001
COMPARE
50/60 230 Rotating Shaker, 230V without Plug
Call For Price
Scientific Industries Bag Rotator Genie Rotating Shaker, 230V with European Plug WW-04724-13 Mfr # SI-1002
COMPARE
50/60 230 Rotating Shaker, 230V with European Plug