Scienceware Tubing adapters, PTFE
For vacuum work or inlet/outlet of gases. Port accepts thermometer, glass tubing, or pipettes. Hose barb connection is of PTFE and accepts 5/16" tubing.
18,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Taper Size
Availability Pricing
Scienceware 20311-1717 Tubing Adapter of PTFE, size 24/40, 8 mm WW-06140-52 Mfr # 20311-1717
COMPARE
24/40
₹18,500.00INR / Each
Scienceware 20311-1718 Tubing Adapter of PTFE, size 24/40, 10 mm WW-06140-53 Mfr # 20311-1718
COMPARE
24/40
₹18,500.00INR / Each