Scienceware Transpette 8-Channel Disposable Transfer Pipettes
Economical, single-use, 8-channel transfer pipettor
  • Ideal for use with samples in micro-wellplates
  • Clear plastic construction
  • 600 µl capacity per channel
Use these multichannel transfer pipetters in place of expensive pipettors and pipette tips. Use to perform plate washing, media changes or supernatant collection. Dispose after "hot" uses to avoid radiation and contamination of other costly pipettors. Not autoclavable.
14,300.00 - ₹67,000.00INR / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile
Availability Pricing
Scienceware Transpette 8-Channel Transfer Pipettors, LDPE, Non-Sterile; 25/Pk WW-07897-06 Mfr # F378730000
COMPARE
No
₹14,300.00INR / Pkg of 25
Scienceware Transpette 8-Channel Transfer Pipettors, LDPE, Sterile; 100/Pk WW-07897-08 Mfr # F378730001
COMPARE
Yes
₹67,000.00INR / Pkg of 100