This product is now discontinued, View Related Products

Scienceware Switch-Grid Tube Racks

25 possible tube size combinations with one rack!
  • Innovative split-frame design holds up to two different-sized tubes in the same rack
  • Interchangeable grid inserts let you quickly and easily change tube sizes
  • Less expensive than purchasing multiple racks
Each rack comes complete with a set of grids in the size of your choice: five color-coded grid inserts fit 10 to 30 mm dia test tubes. The three tiered design holds each tube securely in place while rubber feet keep the rack from sliding on the benchtop and secure stacking while in storage. Constructed of chemical-resistant polyoxymethylene plastic, the frame and grids are easily cleaned with mild detergent, ethanol or isopropyl alcohol. Autoclavable at 250°F (121°C).
6,200.00 - ₹7,700.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Max Tube Size (mm) Tubes Held
Availability Pricing
Scienceware F18745-0010 Switch-Grid Test Tube Rack, Holds 10-13mm Tubes, Orange. WW-06736-12 Mfr # F18745-0010
COMPARE
Orange 13 84
Call For Price
Scienceware F18745-0011 Switch-Grid Test Tube Rack, Holds 13-16mm Tubes, Blue. WW-06736-13 Mfr # F18745-0011
COMPARE
Blue 16 60
Call For Price
Scienceware F18745-0012 Switch-Grid Test Tube Rack, Holds 16-20mm Tubes, Green. WW-06736-14 Mfr # F18745-0012
COMPARE
Green 20 40
₹7,700.00INR / Each
Scienceware F18745-0013 Switch-Grid Test Tube Rack, Holds 20-25mm Tubes, Fuchsia. WW-06736-16 Mfr # F18745-0013
COMPARE
Fuchsia 25 24
₹7,700.00INR / Each
Scienceware F18745-0014 Switch-Grid Test Tube Rack, Holds 25-30mm Tubes, Yellow. WW-06736-17 Mfr # F18745-0014
COMPARE
Yellow 30 12
₹6,200.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat