Scienceware Sterileware Bent Handle Scoops
The curved handle of these scoops elevates the users hand to reduce the possibility of source contamination and exposure to hazardous substances. Made from durable polystyrene they are safe for food, drug, and cosmetic contact. A flat bottom helps keep the scoops upright allowing them to be used as a weight boat. Each individually packaged single use scoop is sterilized by gamma irradiation, lot stamped for accurate tracing, and shipped with certificate of sterilization.
8,600.00 - ₹9,200.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (oz)
Availability Pricing
Scienceware H36902-2006 Sterileware Bent Handle Scoop, 2 oz, 10 pack WW-06263-51 Mfr # H36902-2006
COMPARE
2.028
₹8,600.00INR / Pkg of 10
Scienceware H36904-2006 Sterileware Bent Handle Scoop, 4 oz, 10 pack WW-06263-53 Mfr # H36904-2006
COMPARE
4.226
₹9,200.00INR / Pkg of 10