Scienceware Sterile Red Scoops
Bright red color reduces the chance of scoop being left behind in sample
  • Individually sealed and sterilized in a polythylene bag ready for one time use and disposal
  • Long handle increases distance between hand and sample
  • Remain upright on a flat service, can be used as a weigh boat
  • FDA grade material
Sterilware® scoops are the perfect shape an size for taking samples, especially from wide-mouth containers. Designed to hold various capacities, these one-piece sterile scoops are molded of high-impact polystyrene plastic. Capacities/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
40,100.00 - ₹41,800.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (in) Sterile
Availability Pricing
Scienceware H369022002 Individually Sterile wrapped red scoop - 2 oz., 100/pk WW-06282-60 Mfr # H369022002
COMPARE
60 8 1/16 Yes
₹41,800.00INR / Pkg of 100
Scienceware H369042004 Individually Sterile wrapped red scoop - 4 oz., 100/pk WW-06282-62 Mfr # H369042004
COMPARE
125 8 11/16 Yes
₹40,100.00INR / Pkg of 100