Scienceware SPINRING STIRBARS, PTFE
Removable stabilizing ring keeps stir bar centered in vessel. Octagon shape creates intense vortex without splashing or chattering and resists spinout.
8,500.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Diameter (in)
Availability Pricing
Scienceware F371400010 Spinring Stirring Bar, PTFE, 1-1/4"od WW-04775-42 Mfr # F371400010
COMPARE
0.25
₹8,500.00INR / Each