This product is now discontinued, View Related Products
Scienceware Jerricans, HDPE
Graduated in both gallons and liters, these durable, high-density polyethylene (HDPE) carboys feature a unique 8" (20cm) long spout with a built-in air vent. LDPE spout retracts into the threaded collar to conserve space and is sealed by a leakproof HDPE screw cap. Carboys feature hand grip on the bottom to provide more control when pouring. Large flat molded-in area on sides allow for clear labeling and additional information. Five liter and 10-liter have 3/4" (1.9 cm) ID pouring spout and the 20-liter carboy has a 1" (2.5 cm) ID pouring spout.
11,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Material
Availability Pricing
Scienceware H109350000 5 Liter Jerrican with spout, HDPE WW-06034-04 Mfr # H109350000
COMPARE
5 High Density Polyethyene (HDPE)
₹11,800.00INR / Each
Scienceware H109360000 10 Liter Jerrican with spout, HDPE WW-06034-06 Mfr # H109360000
COMPARE
10 High Density Polyethyene (HDPE)
Call For Price
Scienceware H109370000 20 Liter Jerrican with spout, HDPE WW-06034-08 Mfr # H109370000
COMPARE
20 High Density Polyethyene (HDPE)