This product is now discontinued, View Related Products
Scienceware Earth-Friendly Long Handle Sampling Spoons, PLA
Environmentally conscious sampling
  • FDA compliant spoons are safe for food, drug and cosmetic contact
  • Green alternative to petroleum-based sampling tools
These earth-friendly sampling spoons are durably constructed with PLA (polylactic acid) resin. PLA is a 100% plant-based material which contributes less greenhouse gasses from source to resin than its petroleum-based counterparts. Using annually renewable resources with little environmental impact, these spoons offer a low carbon footprint, are biodegradable, and can be disposed of through industrial composting, incineration, land fill or recycling. They are comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools. Nonsterile.
Long-Handle Spoons are ideal for acquiring samples from bottles, jars, and other confined areas while helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Overall Length (in)
Availability Pricing
Scienceware F36940-1000 Earth-Friendly Long Handle Sampling Spoon, 1/4 tsp, 10 pack WW-06263-87 Mfr # F36940-1000
COMPARE
1.23 Polylactic Acid (PLA) 7 1/8
Call For Price
Scienceware F36942-1000 Earth-Friendly Long Handle Sampling Spoon, 1/2 tsp, 10 pack WW-06263-88 Mfr # F36942-1000
COMPARE
2.46 Polylactic Acid (PLA) 7 1/64
Call For Price
Scienceware F36944-1000 Earth-Friendly Long Handle Sampling Spoon, 1 tsp, 10 pack WW-06263-89 Mfr # F36944-1000
COMPARE
4.93 Polylactic Acid (PLA) 7 1/16