Scienceware Churn Sample Splitters
Mix and divide a water sample into subsamples for testing
  • Polyethylene vessel with polypropylene spigot
  • Swing handle included for ease of transportation
Use the sample splitter to efficiently composite representative cross-section samples of water-sediment mixtures from rivers and streams. Composite samples are homogenized and then split into representative subsamples using the churn splitter. The spring-loaded spigot drains the subsamples into containers for water quality analysis.
199,000.00 - ₹206,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Height (in) Sample Volume
Availability Pricing
Scienceware F37805-0004 4 liter Churn Sample Splitter WW-99160-00 Mfr # F37805-0004
COMPARE
6.5 12 4
₹199,000.00INR / Each
Scienceware F37805-0008 8 liter Churn Sample Splitter WW-99160-02 Mfr # F37805-0008
COMPARE
8.5 11 8
₹206,000.00INR / Each