This product is now discontinued, View Related Products
Sartorius Practum Toploading Balances
Advanced weighting cell with maximum overload protection for years of dependable service
  • Touchscreen swipe technology, enables users to easily navigate through all balance functions
  • Built-in overload protection
  • Supervisor lock to prevent unintentional menu changes
  • Mini USB interface port enables communication with a PC or printer
  • Automatic calibration and adjustment with an external weight
  • Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting, and averaging
282,000.00 - ₹549,000.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Sartorius PRACTUM313-1S Toploading Balance 310g x 0.001g, External Calibration WW-11800-90 Mfr # PRACTUM313-1S
COMPARE
310 100 to 240 0.001
₹429,000.00INR / Each
Sartorius PRACTUM213-1S Toploading Balance 210g x 0.001g, External Calibration WW-11800-91 Mfr # PRACTUM213-1S
COMPARE
210 100 to 240 0.001
₹400,000.00INR / Each
Sartorius PRACTUM2102-1S Toploading Balance 2100g x 0.01g, External Calibration WW-11800-92 Mfr # PRACTUM2102-1S
COMPARE
2100 100 to 240 0.01
₹361,000.00INR / Each
Sartorius PRACTUM1102-1S Topload Balance 1100g x 0.01g, External Calibration WW-11800-93 Mfr # PRACTUM1102-1S
COMPARE
1102 100 to 240 0.01
₹346,000.00INR / Each
Sartorius PRACTUM612-1S Toploading Balance 612, 610g x 0.01g, External Calibration WW-11800-94 Mfr # PRACTUM612-1S
COMPARE
610 100 to 240 0.01
₹315,000.00INR / Each
Sartorius PRACTUM5101-1S Toploading Balance, 5100g x 0.1g, 100-240V WW-11800-95 Mfr # PRACTUM5101-1S
COMPARE
5100 100 to 240 0.1
₹369,000.00INR / Each
Sartorius PRACTUM5100-1S Toploading Balance, 5100g x 1g, 100-240V WW-11800-96 Mfr # PRACTUM5100-1S
COMPARE
5100 100 to 240 1
₹286,000.00INR / Each
Sartorius Practum513-1S Precision Toploading Balance, 510g x 0.001g, 100-240V WW-11955-13 Mfr # PRACTUM513-1S
COMPARE
510 115 0.001
₹549,000.00INR / Each
Sartorius Practum412-1S Toploading Balance 410g x 0.01g, External Calibration WW-11955-14 Mfr # PRACTUM412-1S
COMPARE
410 115 0.01
₹282,000.00INR / Each
Sartorius Practum3102-1S Toploading Balance 3100g x 0.01g, External Calibration WW-11955-15 Mfr # PRACTUM3102-1S
COMPARE
3100 115 0.01
₹429,000.00INR / Each
Sartorius Practum2101-1S Toploading Balance 2100g x 0.1g, External Calibration WW-11955-16 Mfr # PRACTUM2101-1S
COMPARE
2100 115 0.1
₹325,000.00INR / Each
Sartorius Practum6101-1S Toploading Balance 6100g x 0.1g, External Calibration WW-11955-17 Mfr # PRACTUM6101-1S
COMPARE
6100 115 0.1
₹386,000.00INR / Each
Sartorius Practum6100-1S Toploading Balance 6100g x 1.0g, External Calibration WW-11955-18 Mfr # PRACTUM6100-1S
COMPARE
6100 115 1
₹299,000.00INR / Each
Showing 1- 13 of 13