This product is now discontinued, View Related Products
Sartorius Practum Analytical Balances
Advanced weighing cell with maximum overload protection offers years of dependable service
  • High-contrast LCD
  • Built-in overload protection
  • Removable glass draft shield
  • Supervisor lock to prevent unintentional menu changes
  • Mini USB interface enables communication with a PC or printer
  • Automatic calibration and adjustment with an external weight
  • Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting, and averaging
599,000.00 - ₹769,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Sartorius PRACTUM224-1S Analytical Balance, 220g x 0.1mg, External Calibration WW-11800-88 Mfr # PRACTUM224-1S
COMPARE
External 220 0.0001
₹769,000.00INR / Each
Sartorius PRACTUM124-1S Analytical Balance, 120g x 0.1mg, External Calibration WW-11800-89 Mfr # PRACTUM124-1S
COMPARE
External 120 0.0001
₹701,000.00INR / Each
Sartorius Practum64-1S Analytical Balance 60g x 0.1mg, External Calibration WW-11955-12 Mfr # PRACTUM64-1S
COMPARE
External 60 0.0001
₹599,000.00INR / Each