Sartorius Cubis II (MCE) Precision Toploading Balances
Individual configuration of draft shield and integrated ionizer provides adaptation to any weighing task
  • Monolithical weighing system, integrated climate sensors, individual sample holders offer industry leading performance
  • Individual QApp workflows and motorized auto-leveling reduce error
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management provides excellent data integrity
  • Innovative TFT touch screen operation
  • Factory installed set of essential weighing applications
  • Essential user interface (MCE) features high-contrast touch display for easy operation
Toploading balances have a maximum capacity between 320 g and 14,200 g with an easy to use manual glass draft shield. High capacity models have a maximum capacity between 11,200 g and 70,200 g. The flat stainless steel draft shield frame on specific models can be removed. Large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Sartorius Cubis II Precision Balance, 3200 g x 1 mg, with Removable Flat Stainless Steel Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-33 Mfr # MCE3203SR-S00
COMPARE
3200 1 Precision Balance, 3200 g x 1 mg, with Removable Flat Stainless Steel Draft Shield; 100 to 240 VAC, 50/60 Hz
₹2,427,000.00INR / Each
Sartorius Cubis II Precison Balance, 3200 g x 1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-34 Mfr # MCE3203S-2S00-E
COMPARE
3200 1 Precison Balance, 3200 g x 1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz
₹2,427,000.00INR / Each
Sartorius Cubis II Precision Balance, 1200 g x 1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-35 Mfr # MCE1203S-2S00-E
COMPARE
1200 1 Precision Balance, 1200 g x 1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz
₹2,110,000.00INR / Each
Sartorius Cubis II Precision Balance, 620 g x 1 mg, with Removable Flat Stainless Steel Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-36 Mfr # MCE623SR-S00
COMPARE
620 1 Precision Balance, 620 g x 1 mg, with Removable Flat Stainless Steel Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,655,000.00INR / Each
Sartorius Cubis II Precision Balance, 620 g x 1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-37 Mfr # MCE623S-2S00-E
COMPARE
620 1 Precision Balance, 620 g x 1 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,655,000.00INR / Each
Sartorius Cubis II Precison Balance, 8200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-38 Mfr # MCE8201S0-S00
COMPARE
8200 100 Precison Balance, 8200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,222,000.00INR / Each
Sartorius Cubis II Precision Balance, 8200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-39 Mfr # MCE8202S-2S00-0
COMPARE
8200 1 Precision Balance, 8200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,725,000.00INR / Each
Sartorius Cubis II Precision Balance, 6200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-40 Mfr # MCE6202S0-S00
COMPARE
6200 10 Precision Balance, 6200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,524,000.00INR / Each
Sartorius Cubis II Precision Balance, 4200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-41 Mfr # MCE4202S-2S00-0
COMPARE
4200 10 Precision Balance, 4200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,324,000.00INR / Each
Sartorius Cubis II Precision Balance, 2200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-42 Mfr # MCE2202S-2S00-0
COMPARE
2200 10 Precision Balance, 2200 g x 10 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,206,000.00INR / Each
Sartorius Cubis II Precision Balance, 12200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-43 Mfr # MCE12201S-2S00-0
COMPARE
12200 100 Precision Balance, 12200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹1,354,000.00INR / Each
Sartorius Cubis II High Capacity Balance, 70200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-44 Mfr # MCE70201S-2S00-0
COMPARE
70200 100 High Capacity Balance, 70200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹3,670,000.00INR / Each
Sartorius Cubis II High Capacity Balance, 36200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-45 Mfr # MCE36201S0-S00
COMPARE
36200 100 High Capacity Balance, 36200 g x 100 mg, No Draft Shield; 100 to 240 VAC, 50/60 Hz
₹2,280,000.00INR / Each
Showing 1- 13 of 13