Sartorius Cubis II (MCE) Semi Micro Analytical Balances
Individual configuration of draft shield and integrated ionizer provides adaptation to any weighing task
  • Monolithical weighing system, integrated climate sensors, individual sample holders offer industry leading performance
  • Individual QApp workflows and motorized auto-leveling reduce error
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management provides excellent data integrity
  • Innovative TFT touch screen operation
  • Factory installed set of essential weighing applications
  • Activated by touch keypad or touch-free using IR sensor (item 39742-28)
  • Essential user interface (MCE) features high-contrast touch display for easy operation
The Semi-Micro balances have a maximum capacity between 120 g and 520 g with a scale interval of 0.01 mg to 0.1 mg. The automatic glass draft shield (with or without integrated ionizer) removes interfering electrostatic charges on samples and sample containers. The manual glass draft shield has smooth-action doors. Large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sartorius Cubis II Semi Micro Balance, 220 g x 0.01 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz WW-39742-26 Mfr # MCE225SU-S00
COMPARE
Semi Micro Balance, 220 g x 0.01 mg, with Manual Glass Draft Shield; 100 to 240 VAC, 50/60 Hz
₹3,604,000.00INR / Each
Sartorius Cubis II Semi Micro Balance, 220 g x 0.01 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz WW-39742-27 Mfr # MCE225S-2S00-I
COMPARE
Semi Micro Balance, 220 g x 0.01 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz
₹4,610,000.00INR / Each
Sartorius Cubis II Poly Range Semi Mico Balance, 60|120|220 g x 0.01|0.02|0.05 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz WW-39742-28 Mfr # MCE225P-2S00-I
COMPARE
Poly Range Semi Mico Balance, 60|120|220 g x 0.01|0.02|0.05 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz
₹4,114,000.00INR / Each