Sartorius Cubis® II (MCA) Toploading Balances with Software
Long-term flexibility for regulatory and laboratory needs
  • Fully customizable hardware, software, and connectivity
  • Complete data integrity for confident compliance
  • Motorized auto-leveling reduces errors
  • Error-free operation by assessing and correcting relevant factors
  • Electronic signature in compliance with 21 CFR Part 11
  • Safe data transfer
  • Advanced MCA user interface features large color TFT touch screen with swipe functionality
Sartorius® Cubis® II MCA toploading balances are completely customizable to align with your unique application and compliance requirements while ensuring reliable results. The precision balances offer a max capacity between 320 g and 14,200 g with a scale interval range of 1 mg to 100 mg. The high-capacity balances offer a max capacity between 11,200 g and 70200 g with a scale interval range of 100 mg to 1 g. Innovative technology includes automatic motorized leveling, motion controlled and illuminated draft shield, and climate sensor for environmental monitoring to ensure error-free operation.
Large 7” color touch-screen display allows you to touch, scroll, and swipe with factory-installed weighing applications. The status center shows all information about your balance and environmental conditions as well as detailed help and support in case of warnings and errors.
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Sartorius MCA623S-2S00-E Cubis® II  Precision Balance, 620 g x 1 mg; Manual Glass Draft Shield WW-39742-03 Mfr # MCA623S-2S00-E
COMPARE
External 620 1
₹1,872,000.00INR / Each
Sartorius MCA3203S-2S00-E Cubis® II Precision Balance, 3200 g x 1 mg; Manual Glass Draft Shield WW-39742-12 Mfr # MCA3203S-2S00-E
COMPARE
External 3200 1
₹2,643,000.00INR / Each
Sartorius MCA1203S-2S00-E Cubis® II Precision Balance, 1200 g x 1 mg; Manual Glass Draft Shield WW-39742-13 Mfr # MCA1203S-2S00-E
COMPARE
External 1200 1
₹2,327,000.00INR / Each
Sartorius MCA8202S-2S00-0 Cubis® II Precision Balance, 8200 g x 10mg WW-39742-14 Mfr # MCA8202S-2S00-0
COMPARE
External 8200 10
₹1,941,000.00INR / Each
Sartorius MCA6202S-2S00-0 Cubis® II Precision Balance, 6200 g x 10 mg WW-39742-15 Mfr # MCA6202S-2S00-0
COMPARE
External 6200 10
₹1,741,000.00INR / Each
Sartorius MCA4202S-2S00-0 Cubis® II Precision Balance, 4200 g x 10 mg WW-39742-16 Mfr # MCA4202S-2S00-0
COMPARE
External 4200 10
₹1,540,000.00INR / Each
Sartorius MCA2202S-2S00-0 Cubis® II Precision Balance, 2200 g x 10 mg WW-39742-17 Mfr # MCA2202S-2S00-0
COMPARE
External 2200 10
₹1,422,000.00INR / Each
Sartorius MCA8201S-2S00-0 Cubis® II Precision Balance, 8200 g x 100 mg WW-39742-18 Mfr # MCA8201S-2S00-0
COMPARE
External 8200 100
₹1,438,000.00INR / Each
Sartorius MCA12201S-2S00-0 Cubis® II High Capacity Balance, 12,200 g x 100 mg WW-39742-19 Mfr # MCA12201S-2S00-0
COMPARE
External 12200 100
₹1,570,000.00INR / Each
Sartorius MCA70201S-2S00-0 Cubis® II High Capacity Balance, 70,200 g x 100 mg WW-39742-20 Mfr # MCA70201S-2S00-0
COMPARE
External 70200 100
₹3,886,000.00INR / Each
Sartorius MCA36201S-2S00-0 Cubis® II High Capacity Balance, 36,200 g x 100 mg WW-39742-21 Mfr # MCA36201S-2S00-0
COMPARE
External 36200 100
₹2,496,000.00INR / Each
Showing 1- 11 of 11