Sartorius® Cubis® II (MCA) Micro Balance with Software
Long-term flexibility for regulatory and laboratory needs
  • Fully customizable hardware, software, and connectivity
  • Complete data integrity for confident compliance
  • Error-free operation by assessing and correcting relevant factors
  • Advanced MCA user interface with swipe functionality
  • All-inclusive software package included
Sartorius® Cubis® II MCA is a completely customizable balance to align with your unique application and compliance requirements while ensuring reliable results. Innovative technology features draft shield technology, fast and easy motorized leveling, an integrated ionizer for automatic electrostatic elimination, and climate sensor for environmental monitoring to ensure error-free operation.
Large 7” color touch screen display allows you to touch, scroll, and swipe with factory-installed weighing applications. The status center shows all information about your balance and environmental conditions as well as detailed help and support in case of warnings and errors.
The all-inclusive software, QApp 99, includes four different sub-packages for compliance, complex weighing applications, weighing applications and helpful tools, and connectors for data exchange. Full end-to-end data integrity follows ALCOA+ principles. Data cannot be manipulated with the user management function and LIMS connectivity. User management includes centralized password management support, high security management configuration, and login rules. Electronic signatures are in compliance with 21 CFR Part 11.
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Sartorius MCA2.7SM-S00 Cubis® II Ultra Micro Balance, 2.1 g x 0.0001 mg; Auto Glass Draft Shield WW-39742-46 Mfr # MCA2.7SM-S00
COMPARE
Internal 2.1 0.0001
₹10,939,000.00INR / Each
Sartorius MCA6.6SM-S00 Cubis® II Micro Balance, 6.1 g x 0.001 mg; Auto Glass Draft Shield WW-39742-47 Mfr # MCA6.6SM-S00
COMPARE
Internal 6.1 0.001
₹8,732,000.00INR / Each
Sartorius MCA225SI-S00 Cubis® II Semi-Micro Balance, 220 g x 0.01 mg; Auto Glass Draft Shield & Integrated Ionizer WW-39742-48 Mfr # MCA225SI-S00
COMPARE
Internal 220 0.01
₹5,442,000.00INR / Each