This product is now discontinued, View Related Products
Sanitary Mixers
Specifically designed for pharmaceutical, biotech, food and other applications where GMP requirements are standard
  • Compact, rugged, sanitary design with 180-Grit surface finish for wetted surfaces
  • Two dual-sealed radial precision roller bearings for superior shaft support
  • Dry running mechanical seal capable of speeds up to 1200 rpm and pressures from full vacuum to 200 psig
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Max Volume (Liters)
Availability Pricing
Sanitary mixer; stainless steel body, pneumatic direct drive WW-04403-02 Mfr # MXCP-SS-AD
COMPARE
1200 64
Call For Price
Sanitary mixer; stainless steel body, pneumatic gear-reduced drive WW-04403-04 Mfr # MXCP-SS-AG
COMPARE
1200 288
Call For Price
Sanitary mixer; stainless steel body, 115/230 VAC, gear-reduced drive WW-04403-06 Mfr # MXCP-SS-EV
COMPARE
500 352
Call For Price
Sanitary mixer; aluminum body, pneumatic direct drive WW-04403-08 Mfr # MXCP-AL-AD
COMPARE
1200 64
Call For Price
Sanitary mixer; aluminum body, pneumatic gear-reduced drive WW-04403-10 Mfr # MXCP-AL-AG
COMPARE
1200 288
Call For Price
Sanitary mixer; aluminum body, 115/230 VAC, gear-reduced drive WW-04403-12 Mfr # MXCP-AL-EV
COMPARE
500 352