This product is now discontinued, View Related Products

Sampling Spoons, PP

Sampling spoons made of talc-filled polypropylene are autoclavable at 250° (121°C) to maintain sample integrity. Select from various capacities to meet your laboratory needs.
3,400.00 - ₹3,800.00INR / Pkg of 12

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Scienceware F36723-0000 Polypropylene sampling spoon, 1/4 tsp WW-06263-15 Mfr # F36723-0000
COMPARE
Polypropylene sampling spoon, 1/4 tsp
₹3,400.00INR / Pkg of 12
Scienceware F36724-0000 Polypropylene sampling spoon, 1/2 tsp WW-06263-25 Mfr # F36724-0000
COMPARE
Polypropylene sampling spoon, 1/2 tsp
Call For Price
Scienceware F36725-0000 Polypropylene sampling spoon, 1 tsp WW-06263-35 Mfr # F36725-0000
COMPARE
Polypropylene sampling spoon, 1 tsp
₹3,800.00INR / Pkg of 12
Scienceware F36726-0000 Polypropylene sampling spoon, 1 tbsp WW-06263-45 Mfr # F36726-0000
COMPARE
Polypropylene sampling spoon, 1 tbsp
Call For Price
Scienceware F36727-0000 Polypropylene sampling spoon assortment pack WW-06263-55 Mfr # F36727-0000
COMPARE
Polypropylene sampling spoon assortment pack
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat