Bürkle Sampling Control Tape
Labeling and sealing conveniently combined to prevent outflow after sampling
  • Quickly seal holes from sampling lances with these sampling seals
  • Powerful adhesive ensures the seal remains firmly intact
  • Provide quality control data about the sample taken while also sealing where conventional labels will not stick
Sampling control tape is constructed with extremely strong adhesive strength, providing a gas-proof seal. This tape is ideal for use when sealing larger holes or longer tears in containers as it is available on a 164-ft (50m) continuous roll for customizable lengths. This white tape comes in both standard and a food and pharma option.
Food and pharma seals use an adhesive that is approved for use with foodstuff according to FDA guidelines.
Use the 06040-90 dispenser for the 95 x 95 mm labels and the 06040-91 dispenser for the larger 150 x 150 mm labels.
21,200.00 - ₹29,700.00INR / Roll
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
Burkle 5303-0017 Sampling Seal Tape, 95 mm x 50 m WW-06040-67 Mfr # 5303-0017
COMPARE
White 2 3.75
₹21,200.00INR / Roll
Burkle 5303-1017 Sampling Control Seal, 150 x 150 mm; White WW-06040-75 Mfr # 5303-1017
COMPARE
White 6 6
₹29,700.00INR / Roll
Burkle 5303-2019 Sampling Seal Tape, FDA Compliant Adhesive, 95 x 50 m; White WW-06040-78 Mfr # 5303-2019
COMPARE
White 2 3.75
₹21,200.00INR / Roll
Burkle 5303-3019 Sampling Seal Tape, FDA Compliant Adhesive, 150 x 50 m; White WW-06040-85 Mfr # 5303-3019
COMPARE
White 2 5.9
₹29,700.00INR / Roll