BriskHeat® Samox High-Temperature Heating Tapes leads same ends-120 VAC
Get direct-contact heating on conductive surfaces
These heating tapes feature a heavy-duty insulated design, for a maximum exposure temperature of 1400ºF (760ºC). The 120 VAC tapes include a plug and the 240 VAC tapes are supplied with bare leads. The 120 VAC tapes with an opposite-end lead configuration are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC two-prong plug.
16,300.00 - ₹32,900.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat 051100L Samox Heating Tape 1400 and Lt; Deg and Gt; F (760 and Lt; Deg and Gt; C), 1/2" Wide By 10ft Length WW-03106-06 Mfr # TBWH-051-100-LSE
COMPARE
10 1400 120
₹32,900.00INR / Each
BriskHeat 101060L Samox Heating Tape 1400F (760C), 1" Wide By 6ft Length, Leads Same End WW-03106-11 Mfr # TBWH-101-060-LSE
COMPARE
6 1400 120
₹29,600.00INR / Each
BriskHeat BWH051020LSE Samox Insulated Heating Tape Leads On Same End, 2' x 1/2" , 156 Watts, 120 V WW-36115-00 Mfr # BWH051020LSE
COMPARE
2 1400 120
₹16,300.00INR / Each
BriskHeat BWH051-040LSE Samox Insulated Heating Tape Leads On Same End, 4' x 1/2" , 312 Watts, 120 V WW-36115-02 Mfr # BWH051-040LSE
COMPARE
4 1400 120
₹22,000.00INR / Each
BriskHeat BWH051060LSE Samox Insulated Heating Tape Leads On Same End, 6' x 1/2" , 468 Watts, 120 V WW-36115-04 Mfr # BWH051060LSE
COMPARE
6 1400 120
₹22,800.00INR / Each
BriskHeat TBWH051-080-LSE Samox Insulated Heating Tape Leads On Same End, 8' x 1/2" , 624 Watts, 120 V WW-36115-06 Mfr # TBWH051-080-LSE
COMPARE
8 1400 120
₹25,100.00INR / Each
BriskHeat TBWH101-020-LSE Samox Insulated Heating Tape Leads On Same End, 2' x 1" , 314 Watts, 120 V WW-36115-10 Mfr # TBWH101-020-LSE
COMPARE
2 1400 120
₹16,900.00INR / Each
BriskHeat TBWH101-040-LSE Samox Insulated Heating Tape Leads On Same End, 4' x 1" , 628 Watts, 120 V WW-36115-12 Mfr # TBWH101-040-LSE
COMPARE
4 1400 120
₹22,500.00INR / Each
Showing 1- 8 of 8