This product is now discontinued, View Related Products
SalvisLab Coolingcenter Refrigerated Incubators
Precise temperature control for routine lab test or long-term operation
  • Precise control of fan speed from 60 to 100%
  • Storage of up to 50 programs and 15 program steps
  • Temperature regulated by quality PT 100 temperature sensors
SalvisLab Coolingcenter is the perfect refrigerated incubator for every routine test in the lab including high batch operations over long periods. Exclusive EasyMenu microprocessor controller features a digital LCD with easy-to-operate keypad, menu prompts in five selectable languages, intelligent fan speed control, real-time clock, timer, and RS-232 output.
Rugged steel exterior combined with robust insulation minimizes heat/cooling loss, saving energy and keeping exterior temperature at ambient. Mechanical switch on door prevents unit from running when door is open; as soon as it is closed, process will start running again. Over temperature protection shut down operation if max temperature is exceeded.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
SalvisLab Coolingcenter CIC150 Refrigerated Incubator, 150 L, 115 VAC WW-39701-02 Mfr # 31A04176
COMPARE
5.3 115 Refrigerated Incubator, 150 L, 115 VAC
Call For Price
SalvisLab Coolingcenter CIC150 Refrigerated Incubator, 150 L, 220 VAC WW-39701-03 Mfr # 31A04076
COMPARE
5.3 220 Refrigerated Incubator, 150 L, 220 VAC
Call For Price
SalvisLab 31F04210 Wire Shelf for Coolingcenter CIC150 Refrigerated Incubator WW-39701-55 Mfr # 31F04210
COMPARE
Wire Shelf for Coolingcenter CIC150 Refrigerated Incubator