This product is now discontinued, View Related Products

Salvis Incucenter Programmable Incubators

Easy to operate and program!
  • Mechanical forced air convection provides excellent temperature uniformity
  • Natural air convection units are ideal for drying applications that require gentle air flow
Salvis Incucenter programmable mechanical forced-air incubators have the exclusive IntelliFan™ system that provides uniform temperature throughout the chamber independent of load. The maximum temperature of 110°C is ideal for decontamination procedures.

The new Salvis Incucenter programmable natural air convection incubators are ideal for
gentle incubation of organisms, such as on agar plates, and also for conditioning of heat sensitive media.

Programmable capabilities include real-time clock, timer with delayed start from 0 to 999 hours 59 minutes, RS-232 interface, variable fan speed and temperature ramp. LCD display has 5 different languages to choose from for easy programming. Built-in memory back-up retains programs even if power fails! Two separate over-temperature safety features take control of heating and shut down the incubator in the event of main controller failure. The safety glass inner door prevents heat and energy loss while samples are being monitored.

Other features include PT 100 sensor for increased accuracy and a 6 mm access port for additional temperature sensors and cables. The inner chamber is constructed of stainless steel with rounded transitions between walls to make cleaning easy. Purchase optional air filter to reduce contamination in the natural air convection units.
14 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
SalvisLab 31A04140 Programmable Incucenter Incubator, 1.4 cu ft, 115 VAC WW-39701-00 Mfr # 31A04140
COMPARE
1.4 115
Call For Price
SalvisLab 31A04070 Programmable Incucenter Incubator, 1.4 cu ft, 230 VAC WW-39701-05 Mfr # 31A04070
COMPARE
1.4 230
Call For Price
SalvisLab 31A04141 Programmable Incucenter Incubator, 2.8 cu ft, 115 VAC WW-39701-10 Mfr # 31A04141
COMPARE
2.8 115
Call For Price
SalvisLab 31A04071 Programmable Incucenter Incubator, 2.8 cu ft, 230 VAC WW-39701-15 Mfr # 31A04071
COMPARE
2.8 230
Call For Price
SalvisLab 31A04326 Programmable Incucenter Incubator, 5.6 cu ft, 115 VAC WW-39701-20 Mfr # 31A04326
COMPARE
5.6 115
Call For Price
SalvisLab 31A04072 Programmable Incucenter Incubator, 5.6 cu ft, 230 VAC WW-39701-25 Mfr # 31A04072
COMPARE
5.6 230
Call For Price
SalvisLab 31A04143 Programmable Incucenter Incubator, 8.5 cu ft, 115 VAC WW-39701-30 Mfr # 31A04143
COMPARE
8.5 115
Call For Price
SalvisLab 31A04073 Programmable Incucenter Incubator, 8.5 cu ft, 230 VAC WW-39701-35 Mfr # 31A04073
COMPARE
8.5 230
Call For Price
SalvisLab 31A04144 Programmable Incucenter Incubator, 14.0 cu ft, 115 VAC WW-39701-40 Mfr # 31A04144
COMPARE
14 115
Call For Price
SalvisLab 31A04074 Programmable Incucenter Incubator, 14.0 cu ft, 230 VAC WW-39701-45 Mfr # 31A04074
COMPARE
14 230
Call For Price
SalvisLab 31A04160 Programmable Incucenter Natural Convection Incubator, 1.8 cu ft, 115 VAC WW-39702-00 Mfr # 31A04160
COMPARE
1.8 115
₹733,000.00INR / Each
SalvisLab 31A04080 Programmable Incucenter Natural Convection Incubator, 1.8 cu ft, 230 VAC WW-39702-05 Mfr # 31A04080
COMPARE
1.4 230
Call For Price
SalvisLab 31A04161 Programmable Incucenter Natural Convection Incubator, 4.2 cu ft, 115 VAC WW-39702-10 Mfr # 31A04161
COMPARE
2.8 115
Call For Price
SalvisLab 31A04081 Programmable Incucenter Natural Convection Incubator, 4.2 cu ft; 230V WW-39702-15 Mfr # 31A04081
COMPARE
4.2 230
Call For Price
Showing 1- 14 of 14

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat