Salisbury by Honeywell Class 00 Insulated Rubber Gloves
Protect yourself when working with switchboards, power cabinets, or anytime you are working on energized circuits. Combining high dielectric and physical strength, flexibility and durability, gloves provide superior performance meeting and exceeding the requirements of current ASTM D120 specifications and NFPA 70E standards.
* Made of Type I natural rubber
* Meet or exceed ASTM D120 and IEC EN60903
* Available in different colors to allow easy inspection of a glove rotation program
* Glove flexibility makes working with small parts easy
* Glove Inflator (sold separately) simplifies inspection of gloves before each use
19,800.00INR / Pair of 2
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
Salisbury by Honeywell E0011R11 Insulated Glove, Class 00, Red, 11" length, size 11 WW-86432-07 Mfr # E0011R/11
COMPARE
Red 11 11
₹19,800.00INR / Pair of 2