Salisbury by Honeywell Arc Flash Protective Helmet and Face Shield
Tested to ASTM F2178 specifications
* 10 cal/cm2 Arc Thermal Protective Value (ATPV) rating when used with the included chin cup; 8 cal/cm2 without the chin cup
* 7.5" x 20" viewing area
* Extra light tint
* Absorbs >99.9% of harmful UV radiation
* Resistant to fogging
* Tested to ASTM F2178 specifications
62,600.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Salisbury by Honeywell SKA10 Arc Flash Protective Helmet and Face Shield Kit WW-86436-15 Mfr # SKA10
COMPARE
Arc Flash Protective Helmet and Face Shield Kit
₹62,600.00INR / Each