This product is now discontinued, View Related Products
Salisbury by Honeywell Arc Flash Protective Coverall Kits
Take these kits wherever you need arc flash protection, kits come with a convenient carrying bag. Include coveralls, safety glasses, hood, and helmet with face shield
Coverall features
* Made with Indura Ultra Soft® (88% cotton) for comfort and breathability
* Cuff openings and front closure are hook & pile
* 30" (762 mm) inseam with expansion back
* Meet current ASTM F1506 and NFPA 70E standards
* CE 0120 certified
Hood features
* Head and neck protection from arc flash dangers
* Made of two layers of rib knit material with an elastic face opening
Helmet with face shield features
* Face shield has 70% visual light transmission (VLT) — best in the industry!
* 7.5" x 20" viewing area
* Absorbs 99.9% of harmful UV radiation
* Tested to ASTM F2178 specifications
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Size Description
Availability Pricing
Salisbury by Honeywell SKCA8-M Arc Flash Protective Coverall Kits, 8 cal/cm2, medium WW-86436-60 Mfr # SKCA8-M
COMPARE
Medium Arc Flash Protective Coverall Kits, 8 cal/cm2, medium
Call For Price
Salisbury by Honeywell SKCA8-L Arc Flash Protective Coverall Kits, 8 cal/cm2, large WW-86436-61 Mfr # SKCA8-L
COMPARE
Large Arc Flash Protective Coverall Kits, 8 cal/cm2, large
Call For Price
Salisbury by Honeywell SKCA8- Arc Flash Protective Coverall Kits, 8 cal/cm2, extra large WW-86436-62 Mfr # SKCA8-XL
COMPARE
X-Large Arc Flash Protective Coverall Kits, 8 cal/cm2, extra large
Call For Price
Salisbury by Honeywell SKCA8-2 Arc Flash Protective Coverall Kits, 8 cal/cm2, double extra large WW-86436-63 Mfr # SKCA8-2XL
COMPARE
XX-Large Arc Flash Protective Coverall Kits, 8 cal/cm2, double extra large
Call For Price
Salisbury by Honeywell SKCA11-M Arc Flash Protective Coverall Kits, 11 cal/cm2, medium WW-86436-64 Mfr # SKCA11-M
COMPARE
Medium Arc Flash Protective Coverall Kits, 11 cal/cm2, medium
Call For Price
Salisbury by Honeywell SKCA11-L Arc Flash Protective Coverall Kits, 11 cal/cm2, large WW-86436-65 Mfr # SKCA11-L
COMPARE
Large Arc Flash Protective Coverall Kits, 11 cal/cm2, large
Call For Price
Salisbury by Honeywell SKCA11- Arc Flash Protective Coverall Kits, 11 cal/cm2, extra large WW-86436-66 Mfr # SKCA11-XL
COMPARE
X-Large Arc Flash Protective Coverall Kits, 11 cal/cm2, extra large
Call For Price
Salisbury by Honeywell SKCA11-2 Arc Flash Protective Coverall Kits, 11 cal/cm2, double extra large WW-86436-67 Mfr # SKCA11-2XL
COMPARE
XX-Large Arc Flash Protective Coverall Kits, 11 cal/cm2, double extra large
Call For Price
Showing 1- 8 of 8