Justrite Steel Safety Cans for Flammables
Safely contain hazardous liquids and control vapors
  • A necessary part of compliance for safety in the workplace
  • Reduce risk of fire, protecting both personnel and facilities
  • Improve efficiency with ease-of-use features to enhance pouring and filling operations
  • FM approved
  • UL/ULC listed
  • TÜV certified
  • OSHA and NFPA compliant
All cans include a 3 1/2” built-in, stainless-steel flame arrester to prevent flashback ignition. Self-closing, spring loaded lid automatically vents at 3-5 psi (0.2 and 0.35 bar) to fight rupture or explosion. Reinforcing ribs resist denting or implosion. Steel construction with powder coat paint resists corrosion and improves wear and tear. A large ID zone allows you to label content of the can to avoid misuse
13,300.00 - ₹27,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Capacity (Liters) Material
Availability Pricing
Justrite 7110100 Steel Safety Can for Flammables, Type I, Flame Arrester, 1 Gal WW-81791-41 Mfr # 7110100
COMPARE
1 3.79 Steel
₹13,300.00INR / Each
Justrite 7125100 Steel Safety Can for Flammables, Type I, Flame Arrester, 2.5 Gal WW-81791-42 Mfr # 7125100
COMPARE
2.5 9.46 Steel
₹13,700.00INR / Each
Justrite 7150100 Steel Safety Can for Flammables, Type I, Flame Arrester, 5 Gal WW-81791-43 Mfr # 7150100
COMPARE
5 18.93 Steel
₹14,000.00INR / Each
Justrite 7210120 Steel Safety Can for Flammables, Type II, 5/8" Metal Hose, AccuFlow, 1 Gal WW-81791-44 Mfr # 7210120
COMPARE
1 3.79 Steel
₹26,700.00INR / Each
Justrite 7225130 Steel Safety Can for Flammables, Type II, 1" Metal Hose, AccuFlow,  2.5 Gal WW-81791-45 Mfr # 7225130
COMPARE
2.5 9.46 Steel
₹27,200.00INR / Each
Justrite 7250130 Steel Safety Can for Flammables, Type II, 1" Metal Hose, AccuFlow, 5 Gal WW-81791-46 Mfr # 7250130
COMPARE
5 18.93 Steel
₹27,800.00INR / Each