This product is now discontinued, View Related Products

Rubbermaid Versatile Shelving System

A perfect solution for storage and organizing
  • Shelves are steel reinforced and have anti-skid texture
  • Aluminum uprights are non-corrosive
The stationary shelving system comes with 4, 5, or 6 shelves; mobile shelving system is available with four or five shelves. Shelves have a smooth contour and surface for easy cleaning , a perimeter lip, and measure 36" x 20". Optional cabinet kit includes locking two-sided door. Mobile shelves have four 5" dia non-marking swivel casters, two with brakes.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Number of Shelves Shelf Height (in)
Availability Pricing
Rubbermaid 9T36 Stationary shelving unit, 4 shelf, 45" H WW-47209-00 Mfr # 9T36
COMPARE
800 4 45
Call For Price
Rubbermaid 9T37 Stationary shelving unit, 5 shelf, 60" H WW-47209-02 Mfr # 9T37
COMPARE
800 5
Call For Price
Rubbermaid 9T38 Stationary shelving unit, 4 shelf, 72" H WW-47209-04 Mfr # 9T38
COMPARE
800 4 72
Call For Price
Rubbermaid 9T39 Stationary shelving unit, 5 shelf, 72" H WW-47209-06 Mfr # 9T39
COMPARE
800 5
Call For Price
Rubbermaid 9T40 Stationary shelving unit, 6 shelf, 75" H WW-47209-08 Mfr # 9T40
COMPARE
800 6
Call For Price
Rubbermaid 9T42 Mobile Shelving Unit; 5 Shelf, 60"H WW-47209-12 Mfr # 9T42
COMPARE
800 5 60
Call For Price
Rubbermaid 9T43 Mobile Shelving Unit; 4 Shelf, 72"H WW-47209-14 Mfr # 9T43
COMPARE
800 4
Call For Price
Rubbermaid 9T44 Mobile shelving unit, 5 shelf 72" H WW-47209-16 Mfr # 9T44
COMPARE
800 5
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat