This product is now discontinued, View Related Products

Rotronic Disposable Sample Container

Prevents soiling or cross contamination
  • Provides convenient means of collecting and storing samples
The disposable sample containers ensure the optimum sample volume is introduced into the Rotronic WP-14-S sample holders. Prevents sample holders from coming into direct contact with the product being tested, thereby preventing soiling or cross contamination. The sample containers also provide a convenient means of collecting and storing samples.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Overall Height (in) Description
Availability Pricing
Rotromoc E-SCS-00 Replacement Calibration Standard, 0% ERH/; Box of 5 for Water Activity Predictors WW-37910-50 Mfr # E-SCS-00
COMPARE
Replacement Calibration Standard, 0% ERH/; Box of 5 for Water Activity Predictors
Call For Price
Rotromoc E-SCS-10 Replacement Calibration Standard, 10% ERH/; Box of 5, for Use with Water Activity Predictors WW-37910-51 Mfr # E-SCS-10
COMPARE
Replacement Calibration Standard, 10% ERH/; Box of 5, for Use with Water Activity Predictors
Call For Price
Rotromoc EA-35 Replacement Calibration Standard, 35% ERH/; Box of 5, for Water Activity Predictors WW-37910-52 Mfr # EA-35
COMPARE
Replacement Calibration Standard, 35% ERH/; Box of 5, for Water Activity Predictors
Call For Price
Rotromoc EA-80 Replacement Calibration Standard, 80% ERH/; Box of 5, for Water Activity Predictors WW-37910-53 Mfr # EA-80
COMPARE
Replacement Calibration Standard, 80% ERH/; Box of 5, for Water Activity Predictors
Call For Price
Rotromoc PS14 Replacement Sample Cup for Fast Benchtop System, 14 mm WW-37910-67 Mfr # PS14
COMPARE
Transparent 1 3/4 Replacement Sample Cup for Fast Benchtop System, 14 mm
Call For Price
Rotromoc PS40 40mm Replacement Sample Cup for Fast Benchtop System WW-37910-68 Mfr # PS40
COMPARE
40mm Replacement Sample Cup for Fast Benchtop System
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat