This product is now discontinued, View Related Products
Rotational Viscometers
Included 316 stainless steel spindles are ideal for food applications!
  • Select from 10 language options
  • Ensure accurate readings by calibrating your viscometer to a known standard
Determine the dynamic viscosity of samples in applications such as food (sauces, juices, and syrups), adhesives, petroleum products, biofuels, paints, pharmaceuticals, chemicals, and more! Over/under range alarm sounds when your sample is too high/low for the spindle selected. All models feature 10 language options (English, French, German, Italian, Japanese, Portuguese, Spanish, Dutch, Polish, and Catalan), push-button control with easy-to-use menu system, motor self-test, user calibration to a known standard, and universal power supply.
Basic Viscometers are an economical choice for fast, accurate viscosity readings. Feature a 4-line display that shows selected speed, selected spindle, viscosity, and % of full scale.
Basic Viscometers with USB output include software to download data through the USB output to your PC for analysis. Store up to 10 user programs for repeated tests. Display selected speed, selected spindle, viscosity, % torque, shear rate, and shear stress on the 5-line display. Enter the sample density, and the unit calculates and displays kinematic viscosity (cSt) as well.
Advanced Viscometers with Temperature Probe have three times the speed selections and double the speed range of the basic models. Monitor and display temperature with the included RTD temperature probe. Program features include time to torque, time to stop, ramp, and multistep.
Programmable Viscometers feature 2600 speed selections and the highest speed range, up to 250 rpm. Monitor and display temperature with the included RTD temperature probe. Control the viscometer through a PC using the included software; also includes curves, graphs, and database references.
Call For Price
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Cup Number Max Viscosity Range (cP) Power (VAC)
Availability Pricing
Toshniwal Basic Viscometer, 20 to 2,000,000 cP, 100 to 240 VAC, 50/60 Hz WW-15944-40 Mfr # VISCOLEAD ONE L
COMPARE
2000000
Call For Price
Toshniwal Basic Viscometer, 100 to 13,000,000 cP, 100 to 240 VAC, 50/60 Hz WW-15944-41 Mfr # VISCOLEAD ONE R
COMPARE
13000
Call For Price
Toshniwal Basic Viscometer, 200 to 106,000,000 cP, 100 to 240 VAC, 50/60 Hz WW-15944-42 Mfr # VISCOLEAD ONE H
COMPARE
106000000
Call For Price
Toshniwal Viscometer USB 2 Million CP WW-15944-43 Mfr # VISCOLEAD ADV L
COMPARE
2000000
Call For Price
Toshniwal Viscometer USB 13 Million CP WW-15944-44 Mfr # VISCOLEAD ADV R
COMPARE
13000000
Call For Price
Toshniwal Viscometer USB 106 Million CP WW-15944-45 Mfr # VISCOLEAD ADV H
COMPARE
106000000
Call For Price
Toshniwal Viscometer/TEMP 6 Million CP WW-15944-46 Mfr # VISCOLEAD PRO L
COMPARE
6000000
Call For Price
Toshniwal Viscometer/TEMP 40 Million CP WW-15944-47 Mfr # VISCOLEAD PRO R
COMPARE
40000000
Call For Price
Toshniwal Viscometer/TEMP 106 Million CP WW-15944-48 Mfr # VISCOLEAD PRO H
COMPARE
106000000
Call For Price
Toshniwal Program Viscometer 6MILLION CP WW-15944-66 Mfr # PREMIUM L
COMPARE
6000000
Call For Price
Toshniwal Program Viscometer 40000000 CP WW-15944-67 Mfr # PREMIUM R
COMPARE
40000000
Call For Price
Toshniwal Program Viscometer 106000000CP WW-15944-68 Mfr # PREMIUM H
COMPARE
106000000
Call For Price
Showing 1- 12 of 12