RNase AWAY and DNA AWAY

Fast, effective way to remove RNase and DNA contamination
These surface decontaminants provide a safe, fast, and proven alternative to remove nuclease and DNA contamination from benchtops, instruments, pipettors, glass, and plasticware. Conventional methods to remove RNA and DNA contamination use a Diethylpyro-carbonate (DEPC) treatment, which is suspected to be carcinogenic and must be handled with gloves and under approved fume
hoods at all time.
83,700.00 - ₹140,000.00INR / Case of 12

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific RNase Surface Decontaminant, 250 mL Bottles; 12/CS WW-25715-01 Mfr # 7000
COMPARE
RNase Surface Decontaminant, Case of (12) 250 ml bottles
₹140,000.00INR / Case of 12
Thermo Scientific RNase Surface Decontaminant; 475 mL Spray Bottles; 6/CS WW-25715-03 Mfr # 7002
COMPARE
RNase Surface Decontaminant, Case of (6) 475 ml spray bottles
₹83,700.00INR / Case of 6
Thermo Scientific DNA Surface Decontaminant, 250 mL Bottles; 12/CS WW-25715-05 Mfr # 7010
COMPARE
DNA Surface Decontaminant, Case of (12) 250 ml bottles
₹109,000.00INR / Case of 12
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat