This product is now discontinued, View Related Products
Revolutionary Science Pro-Series Incufridge Incubators
Wi-Fi enabled for data logging, remote control and monitoring, and alert notification
  • Program custom protocols for specific times and temperatures
  • Cool, heat, or leave your samples at ambient conditions
  • Check on specimens without compromising the environment—units feature a clear secondary door
  • Feel secure with the two-year warranty, stored settings in case of a power failure, and door lock
These chilling incubators are ideal for water/wastewater, milk, and BOD testing, as well as cell culturing, hybridizations, or storing QC samples. The wide temperature range and rugged, compact design make it a perfect cooling, heating, or ambient temperature incubator for any lab.
Use the large, color touch screen to set and run parameters and programs, or control the unit over Wi-Fi. Log critical data and securely store it in the cloud—all over Wi-Fi, no expensive software needed! Additional features include alert notifications and a door lock to protect valuable samples.
495,000.00 - ₹958,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Revolutionary Science Pro-Series  Incufridge Incubator, 33 L; 120-240 VAC WW-31506-02 Mfr # RS-IF-233PRO
COMPARE
1.2 120 / 240
₹495,000.00INR / Each
Revolutionary Science Pro-Series  Incufridge Incubator, 33 L; 220 VAC WW-31506-04 Mfr # RS-IF-233PRO 220V
COMPARE
1.2 220
Call For Price
Revolutionary Science Pro-Series  Incufridge Incubator, 53 L; 120 VAC WW-31506-06 Mfr # RS-IF-253PRO
COMPARE
1.9 115
₹630,000.00INR / Each
Revolutionary Science Pro-Series  Incufridge Incubator, 53 L; 220 VAC WW-31506-08 Mfr # RS-IF-253PRO 220V
COMPARE
1.9 220
Call For Price
Revolutionary Science Pro-Series  Incufridge Incubator, 73 L; 120 VAC WW-31506-10 Mfr # RS-IF-273PRO
COMPARE
2.6 115
₹958,000.00INR / Each
Revolutionary Science Pro-Series  Incufridge Incubator, 73 L; 220 VAC WW-31506-12 Mfr # RS-IF-273PRO 220V
COMPARE
2.6 220
Call For Price
Revolutionary Science Pro-Series  Incufridge Incubator, 113 L; 120 VAC WW-31506-14 Mfr # RS-IF2113PRO
COMPARE
4 115
Call For Price
Revolutionary Science Pro-Series  Incufridge Incubator, 113 L; 220 VAC WW-31506-16 Mfr # RS-IF-2113PRO 220V
COMPARE
4 220
Call For Price
Showing 1- 8 of 8